Novus Biologicals
Manufacturer Code:NBP213339
Catalog # NBP213339
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: GAAGRLLFGYDSFGNMCGKKNSPVEGAPLSGQDMTLKKHVFFMNSCNLEV KGTQLNRMALCVSNCPEEQLDSLEEVQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Choline transporter-like protein 3; choline transporter-like protein 3 member 3 MGC45474 solute carrier family 44 member 3 UNQ558/PRO1115; Solute carrier family 44 member 3; solute carrier family 44, member 3
Gene Aliases: CTL3; SLC44A3; UNQ558/PRO1115
UniProt ID: (Human) Q8N4M1
Entrez Gene ID: (Human) 126969
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.