Novus Biologicals
Manufacturer Code:NBP191572
Catalog # NBP191572
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC44A3. Peptide sequence TFAILIFFWVLWVAVLLSLGTAGAAQVMEGGQVEYKPLSGIRYMWSYHLI. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Choline transporter-like protein 3; choline transporter-like protein 3 member 3 MGC45474 solute carrier family 44 member 3 UNQ558/PRO1115; Solute carrier family 44 member 3; solute carrier family 44, member 3
Gene Aliases: CTL3; SLC44A3; UNQ558/PRO1115
UniProt ID: (Human) Q8N4M1
Entrez Gene ID: (Human) 126969
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.