Invitrogen
Manufacturer Code:PA569277
Catalog # PIPA569277
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human SLC44A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Choline transporter-like protein 2; CTL2 PP1292 choline transporter-like protein 2; IESCA; Inner ear supporting cell antigen; solute carrier family 44 (choline transporter), member 2; Solute carrier family 44 member 2; solute carrier family 44, member 2; solute carrier family 44, member 2 tv2; testicular tissue protein Li 47
Gene Aliases: 1110028E10Rik; CTL2; PP1292; PSEC0210; RGD1309680; SLC44A2
UniProt ID: (Human) Q8IWA5, (Pig) F1S584, (Bovine) A5D7H3, (Mouse) Q8BY89, (Rat) B4F795, (Guinea pig) Q810F1
Entrez Gene ID: (Horse) 100055038, (Rabbit) 100348956, (Human) 57153, (Pig) 100524730, (Bovine) 617624, (Dog) 484951, (Mouse) 68682, (Rat) 363024, (Guinea pig) 100135617
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.