Novus Biologicals
Manufacturer Code:NBP29458620UL
Catalog # NB023697
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 150-250 of human SLC44A2 (NP_065161.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Choline transporter-like protein 2; CTL2choline transporter-like protein 2 DKFZp666A071 FLJ44586 PP1292 Solute carrier family 44 member 2 solute carrier family 44 member 2; solute carrier family 44 (choline transporter), member 2; Solute carrier family 44 member 2; solute carrier family 44, member 2; testicular tissue protein Li 47
Gene Aliases: CTL2; PP1292; PSEC0210; SLC44A2
UniProt ID: (Human) Q8IWA5
Entrez Gene ID: (Human) 57153
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.