Invitrogen
Manufacturer Code:MA528190
Catalog # PIMA528190
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | VIM-15b |
Immunogen | MV4-11 acute monocyte leukemia cells |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. Store in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD92; CDw92; CDW92 antigen; Choline transporter-like protein 1; solute carrier family 44 (choline transporter), member 1; Solute carrier family 44 member 1; solute carrier family 44, member 1
Gene Aliases: CD92; CDW92; CHTL1; CTL1; SLC44A1
UniProt ID: (Human) Q8WWI5
Entrez Gene ID: (Human) 23446
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.