Proteintech
Manufacturer Code:146871AP150UL
Catalog # 50-172-9642
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC44A1 Fusion Protein Ag6024 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD92; CD92 CDW92 choline transporter-like protein 1 CHTL1 CTL1 SLC44A1; CDw92; CDW92 antigen; Choline transporter-like protein 1; solute carrier family 44 (choline transporter), member 1; Solute carrier family 44 member 1; solute carrier family 44, member 1; transporter-like protein
Gene Aliases: 2210409B22Rik; 4833416H08Rik; AW547365; CD92; CDW92; CHTL1; CTL1; SLC44A1
UniProt ID: (Human) Q8WWI5, (Rat) Q9JJZ7, (Mouse) Q6X893
Entrez Gene ID: (Human) 23446, (Rat) 85254, (Mouse) 100434
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.