Novus Biologicals
Manufacturer Code:NBP182706
Catalog # NBP182706
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:YRRQLERQLQQRQEDDKLFLKINGSSNQEAFV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ23848 large neutral amino acids transporter small subunit 4 LAT4amino acid transporter L-type amino acid transporter 4 MGC34680 Solute carrier family 43 member 2 solute carrier family 43 member 2; L-type amino acid transporter 4; Large neutral amino acids transporter small subunit 4; solute carrier family 43 (amino acid system L transporter), member 2; Solute carrier family 43 member 2
Gene Aliases: LAT4; PP7664; SLC43A2
UniProt ID: (Human) Q8N370
Entrez Gene ID: (Human) 124935
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.