Invitrogen
Manufacturer Code:PA5117339
Catalog # PIPA5117339
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human SLC41A1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MgtE SLC41A1 solute carrier family 41 (magnesium transporter) member 1 solute carrier family 41 member 1 solute carrier family 41 member 1; Solute carrier family 41 member 1
Gene Aliases: AI573938; B230315F01Rik; Slc41a1
UniProt ID: (Mouse) Q8BJA2
Entrez Gene ID: (Mouse) 98396
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.