Invitrogen
Manufacturer Code:PA542409
Catalog # PIPA542409
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC3A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: amino acid transporter 1; amino acid transporter SLC3A1; B(0,+)-type amino acid transport protein; D2; D2h; membrane glycoprotein; NAA-TR; NBAT; Neutral and basic amino acid transport protein rBAT; rBAT; SLC3A1 variant B; SLC3A1 variant C; SLC3A1 variant D; SLC3A1 variant E; SLC3A1 variant F; SLC3A1 variant G; solute carrier family 3 (amino acid transporter heavy chain), member 1; Solute carrier family 3 (cystine dibasic and neutral amino acid transporters activator of cystine dibasic and neutral amino acid transport) member 1 ATR1 CSNU1 D2H FLJ34681 NBAT RBAT; solute carrier family 3 (cystine, dibasic and neutral amino acid transporters), member 1; solute carrier family 3 (cystine, dibasic and neutral amino acid transporters, activator of cystine, dibasic and neutral amino acid transport), member 1; Solute carrier family 3 member 1
Gene Aliases: ATR1; CSNU1; D2H; NBAT; NTAA; RBAT; si:dkey-202m23.2; SLC3A1
UniProt ID: (Human) Q07837, (Mouse) Q91WV7, (Rat) Q64319
Entrez Gene ID: (Rabbit) 100009060, (Dog) 403700, (Horse) 100053537, (Pig) 100144588, (Human) 6519, (Bovine) 535601, (Guinea pig) 100726402, (Mouse) 20532, (Rat) 29484, (Zebrafish) 557757
Molecular Function: amylase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.