Invitrogen
Manufacturer Code:PA5102792
Catalog # PIPA5102792
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLC3A1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: amino acid transporter 1; amino acid transporter 1 ATR1 b(0+)-type amino acid transport protein CSNU1 D2 D2h FLJ34681 NAA-TR NBAT Neutral and basic amino acid transport protein rBAT NTAA RBAT Slc3a1 SLC3A1 variant B SLC3A1 variant C SLC3A1 variant D SLC3A1 variant E SLC3A1 variant F SLC3A1 variant G solute carrier family 3 (amino acid transporter heavy chain) member 1 solute carrier family 3 (cystine dibasic and neutral amino acid transporters) member 1 solute carrier family 3 (cystine dibasic and neutral amino acid transporters activator of cystine dibasic and neutral amino acid transport) member 1 solute carrier family 3 member 1 solute carrier family 3 member 1; B(0,+)-type amino acid transport protein; D2; D2h; NAA-TR; NBAT; Neutral and basic amino acid transport protein rBAT; SLC3A1 variant B; SLC3A1 variant C; SLC3A1 variant D; SLC3A1 variant E; SLC3A1 variant F; SLC3A1 variant G; solute carrier family 3 (amino acid transporter heavy chain), member 1; solute carrier family 3 (cystine, dibasic and neutral amino acid transporters), member 1; solute carrier family 3 (cystine, dibasic and neutral amino acid transporters, activator of cystine, dibasic and neutral amino acid transport), member 1; Solute carrier family 3 member 1
Gene Aliases: ATR1; CSNU1; D2H; NBAT; NTAA; RBAT; SLC3A1
UniProt ID: (Human) Q07837, (Mouse) Q91WV7, (Rat) Q64319
Entrez Gene ID: (Human) 6519, (Mouse) 20532, (Rat) 29484
Molecular Function:
amylase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.