Novus Biologicals
Manufacturer Code:NBP298113UV
Catalog # NB052603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human SLC3A1 fragment. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activator of cystine dibasic and neutral amino acid transport) member 1 amino acid transporter 1 ATR1SLC3A1 variant C B(0+)-type amino acid transport protein CSNU1 D2h D2HSLC3A1 variant D dibasic and neutral amino acid transporters) FLJ34681 member 1 NBATneutral and basic amino acid transport protein rBAT RBATSLC3A1 variant B SLC3A1 variant E SLC3A1 variant F SLC3A1 variant G solute carrier family 3 (cystine dibasic and neutral amino acid transporters; amino acid transporter 1; B(0,+)-type amino acid transport protein; D2h; NBAT; Neutral and basic amino acid transport protein rBAT; SLC3A1 variant B; SLC3A1 variant C; SLC3A1 variant D; SLC3A1 variant E; SLC3A1 variant F; SLC3A1 variant G; solute carrier family 3 (amino acid transporter heavy chain), member 1; solute carrier family 3 (cystine, dibasic and neutral amino acid transporters), member 1; solute carrier family 3 (cystine, dibasic and neutral amino acid transporters, activator of cystine, dibasic and neutral amino acid transport), member 1; Solute carrier family 3 member 1
Gene Aliases: ATR1; CSNU1; D2H; NBAT; RBAT; SLC3A1
UniProt ID: (Human) Q07837
Entrez Gene ID: (Human) 6519
Molecular Function: amylase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.