Abnova
Manufacturer Code:H00006519A01
Catalog # 89-105-457
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 211 a.a. &bsim 320 a.a) partial recombinant protein with GST tag.,SLC3A1 (AAH22386 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant SLC3A1.
For Research Use Only
Protein Aliases: amino acid transporter 1; ATR1; B(0,+)-type amino acid transport protein; CSNU1; D2H; FLJ34681; NBAT; Neutral and basic amino acid transport protein rBAT; RBAT; SLC3A1 variant B; SLC3A1 variant C; SLC3A1 variant D; SLC3A1 variant E; SLC3A1 variant F; SLC3A1 variant G; solute carrier family 3 (amino acid transporter heavy chain), member 1; solute carrier family 3 (cystine, dibasic and neutral amino acid transporters), member 1; solute carrier family 3 (cystine, dibasic and neutral amino acid transporters, activator of cystine, dibasic and neutral amino acid transport), member 1; Solute carrier family 3 member 1
Gene Aliases: ATR1; CSNU1; D2H; NBAT; RBAT; SLC3A1
UniProt ID: (Human) Q07837
Entrez Gene ID: (Human) 6519
Molecular Function: amylase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.