Invitrogen
Manufacturer Code:PA575557
Catalog # PIPA575557
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 150-195 of Human SLC39A9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: solute carrier family 39 (zinc transporter), member 9; Solute carrier family 39 member 9; solute carrier family 39, member 9; zinc transporter SLC39A9; Zinc transporter ZIP9; Zinc transporter ZIP9 Solute carrier family 39 member 9 Zrt- and Irt-like protein 9 ZIP-9 ZIP9 UNQ714/PRO1377; ZIP-9; Zrt- and Irt-like protein 9
Gene Aliases: 2010002A02Rik; 2610511I23Rik; 4833420E20Rik; AW209151; SLC39A9; UNQ714/PRO1377; ZIP-9; ZIP9
UniProt ID: (Human) Q9NUM3, (Mouse) Q8BFU1, (Rat) Q3KR82
Entrez Gene ID: (Human) 55334, (Mouse) 328133, (Rat) 314275
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.