Invitrogen
Manufacturer Code:PA521074
Catalog # PIPA521074
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | An 18 amino acid peptide near the amino terminus of human ZIP9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: solute carrier family 39 (zinc transporter), member 9; Solute carrier family 39 member 9; solute carrier family 39, member 9; UNQ714/PRO1377 ZIP-9 ZIP9 zinc transporter SLC39A9 zinc transporter ZIP9 zrt- and Irt-like protein 9; zinc transporter SLC39A9; Zinc transporter ZIP9; ZIP-9; Zrt- and Irt-like protein 9
Gene Aliases: 2010002A02Rik; 2610511I23Rik; 4833420E20Rik; AW209151; SLC39A9; UNQ714/PRO1377; ZIP-9; ZIP9
UniProt ID: (Human) Q9NUM3, (Mouse) Q8BFU1
Entrez Gene ID: (Human) 55334, (Mouse) 328133
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.