Invitrogen
Manufacturer Code:PA521073
Catalog # PIPA521073
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | An 18 amino acid peptide near the carboxy terminus of human ZIP8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BCG induced integral membrane protein BIGM103; BCG-induced integral membrane protein in monocyte clone 103 protein; LIV-1 subfamily of ZIP zinc transporter 6; LZT-Hs6; Metal cation symporter ZIP8; PP3105 BIGM103 LZT-Hs6 ZIP8 BCG induced integral membrane protein BIGM103 BCG-induced integral membrane protein in monocyte clone 103 protein LIV-1 subfamily of ZIP zinc transporter 6 ZIP-8 Zrt- and Irt-like protein 8 solute carrier family 39 (metal ion transporter) member 8 solute carrier family 39 member 8 zinc transporter ZIP8; solute carrier family 39 (metal ion transporter), member 8; Solute carrier family 39 member 8; ZIP-8; Zrt- and Irt-like protein 8
Gene Aliases: BIGM103; CDG2N; LZT-Hs6; PP3105; SLC39A8; ZIP8
UniProt ID: (Human) Q9C0K1
Entrez Gene ID: (Human) 64116
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.