Novus Biologicals
Manufacturer Code:NBP29484920UL
Catalog # NB024208
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 235-380 of human SLC39A7 (NP_008910.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D6S115E D6S2244EReally interesting new gene 5 protein H2-KE4 HKE4Solute carrier family 39 member 7 HLA class II region expressed gene KE4 KE4 Ke4 gene mouse human homolog of RING5Histidine-rich membrane protein Ke4 solute carrier family 39 (zinc transporter) member 7 zinc transporter SLC39A7 ZIP7; Histidine-rich membrane protein Ke4; HLA class II region expressed gene KE4; Ke4 gene, mouse, human homolog of; Really interesting new gene 5 protein; solute carrier family 39 (zinc transporter), member 7; Solute carrier family 39 member 7; Zinc transporter SLC39A7; ZIP7; Zrt-, Irt-like protein 7
Gene Aliases: D6S115E; D6S2244E; H2-KE4; HKE4; KE4; RING5; SLC39A7; ZIP7
UniProt ID: (Human) Q92504
Entrez Gene ID: (Human) 7922
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.