Invitrogen
Manufacturer Code:PA562893
Catalog # PIPA562893
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLC39A7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DADB-100D22.3 D6S115E D6S2244E H2-KE4 HKE4 KE4 RING5 ZIP7 HLA class II region expressed gene KE4 Ke4 gene mouse human homolog of histidine-rich membrane protein Ke4 really interesting new gene 5 protein solute carrier family 39 member 7 zinc transporter SLC39A7; Histidine-rich membrane protein Ke4; HLA class II region expressed gene KE4; Ke4 gene, mouse, human homolog of; Really interesting new gene 5 protein; solute carrier family 39 (zinc transporter), member 7; Solute carrier family 39 member 7; Zinc transporter SLC39A7; ZIP7; Zrt-, Irt-like protein 7
Gene Aliases: D6S115E; D6S2244E; H2-KE4; HKE4; KE4; RING5; SLC39A7; ZIP7
UniProt ID: (Human) Q92504
Entrez Gene ID: (Human) 7922
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.