Invitrogen
Manufacturer Code:PA542523
Catalog # PIPA542523
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC39A5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Solute carrier family 39 (metal ion transporter) member 5 LZT-Hs7 MGC34778 ZIP5; solute carrier family 39 (metal ion transporter), member 5; solute carrier family 39 (zinc transporter), member 5; Solute carrier family 39 member 5; Zinc transporter ZIP5; ZIP-5; Zrt- and Irt-like protein 5
Gene Aliases: 1810013D05Rik; 2010205A06Rik; LZT-Hs7; MYP24; SLC39A5; ZIP5
UniProt ID: (Human) Q6ZMH5, (Mouse) Q9D856
Entrez Gene ID: (Horse) 100052215, (Dog) 481108, (Human) 283375, (Bovine) 511782, (Rabbit) 100343963, (Guinea pig) 100720549, (Rat) 362812, (Mouse) 72002
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.