Novus Biologicals
Manufacturer Code:NBP294856100UL
Catalog # NB024221
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 23-327 of human SLC39A4 (NP_570901.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acrodermatitis enteropathica zinc-deficiency type FLJ20327 MGC74741 solute carrier family 39 (zinc transporter) member 4 Solute carrier family 39 member 4 zinc transporter ZIP4 ZIP-4 ZIP4AEZ Zrt- and Irt-like protein 4; solute carrier family 39 (zinc transporter), member 4; Solute carrier family 39 member 4; Zinc transporter ZIP4; ZIP-4; Zrt- and Irt-like protein 4
Gene Aliases: AEZ; AWMS2; SLC39A4; ZIP4
UniProt ID: (Human) Q6P5W5
Entrez Gene ID: (Human) 55630
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.