Invitrogen
Manufacturer Code:PA578528
Catalog # PIPA578528
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human SLC39A4. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AEZ AWMS2 ZIP4 solute carrier family 39 (zinc transporter) member 4; solute carrier family 39 (zinc transporter), member 4; Solute carrier family 39 member 4; Zinc transporter ZIP4; ZIP-4; Zrt- and Irt-like protein 4
Gene Aliases: AEZ; AWMS2; SLC39A4; ZIP4
UniProt ID: (Human) Q6P5W5
Entrez Gene ID: (Human) 55630
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.