Invitrogen
Manufacturer Code:PA547915
Catalog # PIPA547915
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse SLC39A4 Met1-Tyr337 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Activated in W/Wv mouse stomach 2; AEZ AWMS2 ZIP4 ZIP-4 solute carrier family 39 member 4 zinc transporter ZIP4 zrt- and Irt-like protein 4; mAWMS2; Solute carrier family 39 member 4; Zinc transporter ZIP4; ZIP-4; Zrt- and Irt-like protein 4
Gene Aliases: 1600025H15Rik; AU041686; AWMS2; Slc39a4; ZIP4
UniProt ID: (Mouse) Q78IQ7
Entrez Gene ID: (Mouse) 72027
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.