Invitrogen
Manufacturer Code:PA5113024
Catalog # PIPA5113024
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Zinc transporter ZIP4 protein (23-327AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1600025H15Rik Activated in W/Wv mouse stomach 2 AEZ AU041686 AWMS2 EMBL:AAI27515.1} mAWMS2 Slc39a4 slc39a4 {ECO:0000312 solute carrier family 39 (zinc transporter) member 4 Solute carrier family 39 member 4 Zinc transporter ZIP4 Zip4 ZIP-4 Zrt- and Irt-like protein 4; Activated in W/Wv mouse stomach 2; mAWMS2; solute carrier family 39 (zinc transporter), member 4; Solute carrier family 39 member 4; Zinc transporter ZIP4; ZIP-4; Zrt- and Irt-like protein 4
Gene Aliases: 1600025H15Rik; AEZ; AU041686; AWMS2; SLC39A4; ZIP4
UniProt ID: (Human) Q6P5W5, (Mouse) Q78IQ7
Entrez Gene ID: (Human) 55630, (Mouse) 72027
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.