Invitrogen
Manufacturer Code:PA563582
Catalog # PIPA563582
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLC39A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: solute carrier family 39 (zinc transporter), member 3; Solute carrier family 39 member 3; Zinc transporter ZIP3; ZIP-3; ZIP3 ZIP-3 solute carrier family 39 member 3 zinc transporter ZIP3 zrt- and Irt-like protein 3; Zrt- and Irt-like protein 3
Gene Aliases: SLC39A3; ZIP-3; ZIP3
UniProt ID: (Human) Q9BRY0
Entrez Gene ID: (Human) 29985
Molecular Function:
transmembrane receptor regulatory/adaptor protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.