Novus Biologicals
Manufacturer Code:NBP181551
Catalog # NBP181551
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:EHHHGHSHYASESLPSKKDQEEGVMEKLQNGDLDHMIPQHCSSELDGKAPMVDEKVIVGSLSVQDLQASQSACYWLKGVRYSDIGTLAWMITLSD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Irt-like protein 14 LIV-1 subfamily of ZIP zinc transporter 4 LZT-Hs4 solute carrier family 39 (metal ion transporter) member 14 solute carrier family 39 (zinc transporter) member 14 solute carrier family 39 member 14 zinc transporter ZIP14 ZIP14 ZIP-14 zrt- and Irt-like protein 14; LIV-1 subfamily of ZIP zinc transporter 4; LZT-Hs4; Metal cation symporter ZIP14; solute carrier family 39 (metal ion transporter), member 14; solute carrier family 39 (zinc transporter), member 14; Solute carrier family 39 member 14; ZIP-14; Zrt- and Irt-like protein 14; Zrt-, Irt-like protein 14
Gene Aliases: cig19; HMNDYT2; KIAA0062; LZT-Hs4; NET34; SLC39A14; ZIP14
UniProt ID: (Human) Q15043
Entrez Gene ID: (Human) 23516
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.