Novus Biologicals
Manufacturer Code:NBP176509
Catalog # NBP176509
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human ZIP13. The immunogen is located within amino acids 150 - 200 of ZIP13. Amino Acid Squence: DSKEEKTSQAPNKD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ25785 LIV-1 subfamily of ZIP zinc transporter 9 LZT-Hs9 solute carrier family 39 (metal ion transporter) member 13 solute carrier family 39 (zinc transporter) member 13 Solute carrier family 39 member 13 zinc transporter ZIP13 ZIP13 ZIP-13 Zrt- and Irt-like protein 13; LIV-1 subfamily of ZIP zinc transporter 9; LZT-Hs9; solute carrier family 39 (metal ion transporter), member 13; solute carrier family 39 (zinc transporter), member 13; Solute carrier family 39 member 13; Zinc transporter ZIP13; ZIP-13; Zrt- and Irt-like protein 13
Gene Aliases: LZT-Hs9; SLC39A13; ZIP13
UniProt ID: (Human) Q96H72
Entrez Gene ID: (Human) 91252
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.