Abnova
Manufacturer Code:PAB19372
Catalog # 89-120-906
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 15 amino acids near N-terminus of human SLC39A12. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SLC39A12.
For Research Use Only
Protein Aliases: bA570F3.1; FLJ30499; LIV-1 subfamily of ZIP zinc transporter 8; LZT-Hs8; MGC43205; MGC51099; solute carrier family 39 (metal ion transporter), member 12; solute carrier family 39 (zinc transporter), member 12; Solute carrier family 39 member 12; Zinc transporter ZIP12; ZIP-12; Zrt- and Irt-like protein 12
Gene Aliases: bA570F3.1; LZT-Hs8; SLC39A12; ZIP-12; ZIP12
UniProt ID: (Human) Q504Y0
Entrez Gene ID: (Human) 221074
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.