Abnova
Manufacturer Code:H00057181A01
Catalog # 89-107-177
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 514 a.a. &bsim 621 a.a) partial recombinant protein with GST tag.,SLC39A10 (NP_065075 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant SLC39A10.
For Research Use Only
Protein Aliases: DKFZp781L10106; LZT-Hs2; MGC126565; MGC138428; solute carrier family 39 (metal ion transporter), member 10; solute carrier family 39 (zinc transporter), member 10; Solute carrier family 39 member 10; Zinc transporter ZIP10; ZIP-10; Zrt- and Irt-like protein 10
Gene Aliases: KIAA1265; LZT-Hs2; SLC39A10; ZIP10
UniProt ID: (Human) Q9ULF5
Entrez Gene ID: (Human) 57181
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.