Abnova
Manufacturer Code:PAB19346
Catalog # 89-120-879
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 16 amino acids near N-terminus of human SLC39A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody rasied against synthetic peptide of SLC39A1.
For Research Use Only
Protein Aliases: solute carrier family 39 (zinc transporter), member 1; solute carrier family 39 (zinc transporter), member 3; Solute carrier family 39 member 1; Zinc transporter ZIP1; Zinc-iron-regulated transporter-like; ZIP-1; ZIP1; ZIRTL; Zrt- and Irt-like protein 1
Gene Aliases: CGI-08; CGI-71; IRT1; SLC39A1; ZIP1; ZIRTL
UniProt ID: (Human) Q9Y380
Entrez Gene ID: (Human) 27173
Molecular Function: transmembrane receptor regulatory/adaptor protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.