Invitrogen
Manufacturer Code:PA542504
Catalog # PIPA542504
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC38A5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: amino acid transport system N2; amino acid transporter system N2; Sodium-coupled neutral amino acid transporter 5; solute carrier family 38 (amino acid transporter), member 5; Solute carrier family 38 member 5; Solute carrier family 38 member 5 JM24 SN2 pp7194 SNAT5; solute carrier family 38, member 5; System N transporter 2; transport system N, protein 2
Gene Aliases: C81234; E330031E14; JM24; PP7194; SLC38A5; SN2; SNAT5
UniProt ID: (Human) Q8WUX1, (Bovine) Q5E9S9, (Mouse) Q8BW30, (Rat) A2VCW5
Entrez Gene ID: (Human) 92745, (Bovine) 512495, (Rabbit) 100302413, (Dog) 612527, (Pig) 100627516, (Horse) 100051531, (Mouse) 209837, (Rat) 192208
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.