Abnova
Manufacturer Code:PAB24534
Catalog # 89-126-251
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SLC38A5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant SLC38A5.
For Research Use Only
Protein Aliases: JM24; pp7194; SN2; Sodium-coupled neutral amino acid transporter 5; solute carrier family 38 (amino acid transporter), member 5; Solute carrier family 38 member 5; solute carrier family 38, member 5; System N transporter 2; transport system N, protein 2
Gene Aliases: JM24; PP7194; SLC38A5; SN2; SNAT5
UniProt ID: (Human) Q8WUX1
Entrez Gene ID: (Human) 92745
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.