Invitrogen
Manufacturer Code:PA542143
Catalog # PIPA542143
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC38A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mNAT; N system amino acids transporter NAT-1; N-system amino acid transporter 1; Na(+)-coupled neutral amino acid transporter 3; SLC38A3; slc38a3 {ECO:0000312|RGD:628620}; Sodium-coupled neutral amino acid transporter 3; Solute carrier family 38 member 3; Solute carrier family 38 member 3 G17 SN1 NAT1; solute carrier family 38, member 3; System N amino acid transporter 1; system N1 amino acid transporter; system N1 Na+ and H+-coupled glutamine transporter
Gene Aliases: 0610012J02Rik; D9Ucla2; G17; mNAT; NAT1; Slc38-3; SLC38A3; SN1; SNAT3
UniProt ID: (Human) Q99624, (Rat) Q9JHZ9, (Mouse) Q9DCP2
Entrez Gene ID: (Dog) 476617, (Human) 10991, (Rabbit) 100339311, (Pig) 100156039, (Horse) 100062146, (Sheep) 101116182, (Bovine) 326576, (Guinea pig) 100715348, (Rat) 252919, (Mouse) 76257
Molecular Function:
amino acid transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.