Invitrogen
Manufacturer Code:PA542419
Catalog # PIPA542419
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC38A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Amino acid transporter A1; amino acid transporter system A1; Glutamine transporter; N-system amino acid transporter 2; rATA1; Sodium-coupled neutral amino acid transporter 1; Solute carrier family 38 member 1; Solute carrier family 38 member 1 ATA1 NAT2 SAT1 SNAT1; solute carrier family 38, member 1; System A amino acid transporter 1; System A transporter 2; System N amino acid transporter 1
Gene Aliases: ATA1; GlnT; NAT2; Sa2; SAT1; SLC38A1; SNAT1
UniProt ID: (Human) Q9H2H9, (Rat) Q9JM15
Entrez Gene ID: (Dog) 477633, (Horse) 100050256, (Human) 81539, (Rat) 170567, (Guinea pig) 100725601
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.