Invitrogen
Manufacturer Code:PA5117341
Catalog # PIPA5117341
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human SLC38A1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA408026 AA409865 AL022800 Amino acid transporter A1 amino acid transporter system A1 ATA1 AU015942 GlnT Glutamine transporter MNat2 Nat2 N-system amino acid transporter 2 rATA1 S38A1 Sa2 SAT1 SLC38A1 SNAT 1 Snat1 Sodium-coupled neutral amino acid transporter 1 solute carrier family 38 member 1 solute carrier family 38 member 1 system A amino acid transporter 1 System A transporter 2 system N amino acid transporter 1; Amino acid transporter A1; amino acid transporter system A1; N-system amino acid transporter 2; Sodium-coupled neutral amino acid transporter 1; Solute carrier family 38 member 1; solute carrier family 38, member 1; System A amino acid transporter 1; System N amino acid transporter 1
Gene Aliases: ATA1; NAT2; SAT1; SLC38A1; SNAT1
UniProt ID: (Human) Q9H2H9
Entrez Gene ID: (Human) 81539
Molecular Function:
amino acid transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.