Invitrogen
Manufacturer Code:PA558599
Catalog # PIPA558599
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLC37A4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucose-5-phosphate transporter; glucose-6-phosphatase, transport (glucose) protein 3; glucose-6-phosphatase, transport (glucose-6-phosphate) protein 1; glucose-6-phosphatase, transport (phosphate/pyrophosphate) protein 2; Glucose-6-phosphate exchanger SLC37A4; Glucose-6-phosphate translocase; microsomal glucose-6-phosphate transporter; Solute carrier family 37 (glucose-6-phosphate transporter) member 4 G6PT1 G6PT2 G6PT3 GSD1b GSD1c GSD1d MGC15729 PRO0685 TRG19 TRG-19; solute carrier family 37 (glucose-6-phosphate transporter), member 4; Solute carrier family 37 member 4; Transformation-related gene 19 protein; TRG-19
Gene Aliases: G6PT; G6PT1; G6PT2; G6PT3; GSD1b; GSD1c; GSD1d; PRO0685; SLC37A4; TRG-19; TRG19
UniProt ID: (Human) O43826
Entrez Gene ID: (Human) 2542
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.