Invitrogen
Manufacturer Code:PA542500
Catalog # PIPA542500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC37A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: solute carrier family 37 (glycerol-3-phosphate transporter), member 3; Solute carrier family 37 member 3; solute carrier family 37, member 3; Sugar phosphate exchanger 3; sugar phosphate exchanger 3-like protein
Gene Aliases: SLC37A3; SPX3
UniProt ID: (Bovine) Q17QZ3, (Human) Q8NCC5
Entrez Gene ID: (Bovine) 506910, (Human) 84255, (Horse) 100065965, (Dog) 482768, (Rabbit) 103345720, (Guinea pig) 100718612, (Rat) 312255
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.