Novus Biologicals
Manufacturer Code:NBP257417
Catalog # NBP257417
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VSPEEIGLSGIEAEENFEEDSHRPLINGGE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp761N0624 MGC32939 solute carrier family 37 (glycerol-3-phosphate transporter) member 3 Solute carrier family 37 member 3 SPX3 sugar phosphate exchanger 3; solute carrier family 37 (glycerol-3-phosphate transporter), member 3; Solute carrier family 37 member 3; solute carrier family 37, member 3; Sugar phosphate exchanger 3
Gene Aliases: SLC37A3; SPX3
UniProt ID: (Human) Q8NCC5
Entrez Gene ID: (Human) 84255
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.