Abnova
Manufacturer Code:H00084255K
Catalog # 89-207-751
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SLC37A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SLC37A3 peptide using ARM Technology.
For Research Use Only
Protein Aliases: MGC32939; solute carrier family 37 (glycerol-3-phosphate transporter), member 3; Solute carrier family 37 member 3; solute carrier family 37, member 3; Sugar phosphate exchanger 3
Gene Aliases: SLC37A3; SPX3
UniProt ID: (Human) Q8NCC5
Entrez Gene ID: (Human) 84255
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.