Invitrogen
Manufacturer Code:PA5113138
Catalog # PIPA5113138
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Glucose-6-phosphate exchanger SLC37A2 protein (46-84AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cAMP inducible 2 protein; cAMP inducible 2 protein cAMP inducible gene 2 cAMP-inducible protein 2 Ci2 cI-2 G3PP Glucose-6-phosphate exchanger SLC37A2 pp11662 RGD1564160 Slc37a1 SLC37A2 solute carrier family 37 (glucose-6-phosphate transporter) member 2 solute carrier family 37 (glycerol-3-phosphate transporter) member 1 solute carrier family 37 (glycerol-3-phosphate transporter) member 2 Solute carrier family 37 member 2 sugar phosphate exchanger 2; cAMP inducible gene 2; cAMP-inducible protein 2; Glucose-6-phosphate exchanger SLC37A2; solute carrier family 37 (glucose-6-phosphate transporter), member 2; solute carrier family 37 (glycerol-3-phosphate transporter), member 1; solute carrier family 37 (glycerol-3-phosphate transporter), member 2; Solute carrier family 37 member 2; sugar phosphate exchanger 2
Gene Aliases: cI-2; Ci2; G3PP; pp11662; Slc37a1; SLC37A2
UniProt ID: (Human) Q8TED4, (Mouse) Q9WU81
Entrez Gene ID: (Human) 219855, (Mouse) 56857
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.