Invitrogen
Manufacturer Code:PA5107265
Catalog # PIPA5107265
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide located within the following region: FSLCLLFAKL VSYTFLYWLP LYIFNVAHFS AKEAGDLSTL FDVGGIIGGI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cAMP inducible 2 protein; cAMP inducible gene 2; cAMP-inducible protein 2; Glucose-6-phosphate exchanger SLC37A2; solute carrier family 37 (glycerol-3-phosphate transporter), member 1; Solute carrier family 37 member 2; sugar phosphate exchanger 2
Gene Aliases: cI-2; Ci2; G3PP; Slc37a1; Slc37a2
UniProt ID: (Mouse) Q9WU81
Entrez Gene ID: (Mouse) 56857
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.