Abnova
Manufacturer Code:PAB19213
Catalog # 89-120-743
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 15 amino acids near internal region of human SLC37A2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody rasied against synthetic peptide of SLC37A2.
For Research Use Only
Protein Aliases: FLJ00171; Glucose-6-phosphate exchanger SLC37A2; MGC71430; pp11662; solute carrier family 37 (glucose-6-phosphate transporter), member 2; solute carrier family 37 (glycerol-3-phosphate transporter), member 2; Solute carrier family 37 member 2; sugar phosphate exchanger 2
Gene Aliases: pp11662; SLC37A2
UniProt ID: (Human) Q8TED4
Entrez Gene ID: (Human) 219855
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.