Proteintech
Manufacturer Code:245821AP150UL
Catalog # 50-173-4436
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC37A1 Fusion Protein Ag20180 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G 3 P permease G 3 P transporter G3PP Glycerol 3 phosphate permease SLC37A1; G-3-P permease; G-3-P transporter; Glucose-6-phosphate exchanger SLC37A1; Glycerol-3-phosphate permease; glycerol-3-phosphate transporter; solute carrier family 37 (glucose-6-phosphate transporter), member 1; solute carrier family 37 (glycerol-3-phosphate transporter), member 1; Solute carrier family 37 member 1
Gene Aliases: G3PP; SLC37A1
UniProt ID: (Human) P57057, (Mouse) Q8R070
Entrez Gene ID: (Human) 54020, (Mouse) 224674
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.