Novus Biologicals
Manufacturer Code:NBP250237B
Catalog # NBP250237B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | PAT-4/9/H10 |
Immunogen | Antigenic amino acid sequence within the N-terminus of PAT4 (REELDMDVMRPLINE-C). |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Proton-coupled amino acid transporter 4; Proton/amino acid transporter 4; proton/amino acid transporter 4 solute carrier family 36 (proton/amino acid symporter) member 4; solute carrier family 36 (proton/amino acid symporter), member 4; Solute carrier family 36 member 4
Gene Aliases: PAT4; SLC36A4
UniProt ID: (Human) Q6YBV0
Entrez Gene ID: (Human) 120103
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.