Invitrogen
Manufacturer Code:PA542528
Catalog # PIPA542528
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC36A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Proton-coupled amino acid transporter 3; Proton/amino acid transporter 3; solute carrier family 36 (proton/amino acid symporter), member 3; Solute carrier family 36 member 3; solute carrier family 36, member 3; tramdorin 2; Tramdorin-2
Gene Aliases: PAT3; SLC36A3; TRAMD2; tramdorin2
UniProt ID: (Human) Q495N2
Entrez Gene ID: (Human) 285641
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.