Abnova
Manufacturer Code:H00285641K
Catalog # 89-207-651
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SLC36A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SLC36A3 peptide using ARM Technology.
For Research Use Only
Protein Aliases: FLJ16658 MGC119638 MGC119639 MGC119641 PAT3 TRAMD2 tramdorin2; Proton-coupled amino acid transporter 3; Proton/amino acid transporter 3; solute carrier family 36 (proton/amino acid symporter), member 3; Solute carrier family 36 member 3; solute carrier family 36, member 3; tramdorin 2; Tramdorin-2
Gene Aliases: PAT3; SLC36A3; TRAMD2; tramdorin2
UniProt ID: (Human) Q495N2
Entrez Gene ID: (Human) 285641
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.