Abnova
Manufacturer Code:H00153201K
Catalog # 89-204-755
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SLC36A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SLC36A2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: FLJ16051 MGC119658 MGC119660 PAT2 TRAMD1; Proton-coupled amino acid transporter 2; Proton/amino acid transporter 2; solute carrier family 36 (proton/amino acid symporter), member 2; Solute carrier family 36 member 2; Tramdorin-1
Gene Aliases: PAT2; SLC36A2; TRAMD1
UniProt ID: (Human) Q495M3
Entrez Gene ID: (Human) 153201
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.