Novus Biologicals
Manufacturer Code:NBP230037
Catalog # NBP230037
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This SLC36A1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human SLC36A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: lysosomal amino acid transporter 1; PAT1 solute carrier family 36 (proton/amino acid symporter) member 1; Proton-coupled amino acid transporter 1; Proton/amino acid transporter 1; solute carrier family 36 (proton/amino acid symporter), member 1; Solute carrier family 36 member 1
Gene Aliases: Dct1; LYAAT1; PAT1; SLC36A1; TRAMD3
UniProt ID: (Human) Q7Z2H8
Entrez Gene ID: (Human) 206358
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.