Novus Biologicals
Manufacturer Code:NBP297679JF646
Catalog # NB046461
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human SLC35E3 fragment. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bladder cancer overexpressed protein; Bladder Cancer Overexpressed Protein Bladder Cancer-Overexpressed Gene 1 Protein BLOV1 solute carrier family 35 member E2 Solute Carrier Family 35 Member E3; Bladder cancer-overexpressed gene 1 protein; Solute carrier family 35 member E3; solute carrier family 35, member E2; solute carrier family 35, member E3
Gene Aliases: BLOV1; SLC35E3; UNQ3043/PRO9859
UniProt ID: (Human) Q7Z769
Entrez Gene ID: (Human) 55508
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.