Invitrogen
Manufacturer Code:PA572721
Catalog # PIPA572721
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC35D3 antibody for a 17 amino acid synthetic peptide near the carboxy terminus of human SLC35D3. The immunogen is within the last 50 amino acids of SLC35D3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: frc, fringe-like 1; FRCL1; Fringe connection-like protein 1; Solute carrier family 35 member D3; solute carrier family 35, member D3
Gene Aliases: 6230421J19Rik; FRCL1; SLC35D3
UniProt ID: (Human) Q5M8T2, (Mouse) Q8BGF8
Entrez Gene ID: (Human) 340146, (Mouse) 76157
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.