Novus Biologicals
Manufacturer Code:NBP29445120UL
Catalog # NB023451
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 310-390 of human SLC35D3 (NP_001008783.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA55K22.3 frc fringe-like 1 FRCL1 Fringe connection-like protein 1 MGC102873 solute carrier family 35 member D3 solute carrier family 35 member D3; frc, fringe-like 1; Fringe connection-like protein 1; Solute carrier family 35 member D3; solute carrier family 35, member D3
Gene Aliases: FRCL1; SLC35D3
UniProt ID: (Human) Q5M8T2
Entrez Gene ID: (Human) 340146
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.