Invitrogen
Manufacturer Code:PA572720
Catalog # PIPA572720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC35D2 antibody for a 14 amino acid synthetic peptide near the carboxy terminus of human SLC35D2. The immunogen is within the last 50 amino acids of SLC35D2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: fringe connection; HFRC1; Homolog of Fringe connection protein 1; solute carrier family 35 (UDP-GlcNAc/UDP-glucose transporter), member D2; Solute carrier family 35 member D2; solute carrier family 35, member D2; SQV7-like protein; SQV7L; UDP-galactose transporter-related protein 8; UDP-N-acetylglucosamine transporter; UDP-N-acetylglucosamine/UDP-glucose/GDP-mannose transporter; UGTrel8
Gene Aliases: HFRC; HFRC1; SLC35D2; SQV7L; UGTREL8
UniProt ID: (Human) Q75W21
Entrez Gene ID: (Human) 11046
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.